Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SH3KBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142004
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142004 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142004 Supplier Invitrogen™ Supplier No. PA5142004
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize isoform a (NP_114098.1) only. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:4,000.

This gene encodes an adapter protein that contains three N-terminal Src homology domains, a proline rich region and a C-terminal coiled-coil domain. The encoded protein facilitates protein-protein interactions and has been implicated in numerous cellular processes including apoptosis, cytoskeletal rearrangement, cell adhesion and in the regulation of clathrin-dependent endocytosis. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3KBP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Sh3kbp1
Gene Accession No. Q96B97
Gene Alias 1200007H22Rik; 1700125L08Rik; 5830464D22Rik; AI447724; cbl-interacting protein of 85 kDa; c-Cbl-interacting protein; CD2-binding protein 3; CD2BP3; CIN85; GIG10; HSB1; HSB-1; Human Src family kinase-binding protein 1; IN85; LOW QUALITY PROTEIN: SH3 domain-containing kinase-binding protein 1; MIG18; migration-inducing gene 18; regulator of ubiquitous kinase; Ruk; Seta; SH3 domain containing kinase binding protein 1; SH3 domain-containing adapter protein; SH3 domain-containing kinase-binding protein 1; SH3-containing, expressed in tumorigenic astrocytes; SH3-domain kinase binding protein 1; Sh3kbp1; Src family kinase-binding protein 1; src-related kinase binding protein-1
Gene Symbols Sh3kbp1
Host Species Goat
Immunogen Peptide with sequence VEAIVEFDYQAQH-C (aa 2-14).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30011
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.