Description
- Electrostatically attract tissue sections and cytology preparations, binding them to the slide
- Eliminate the need for special adhesives or protein coatings
- No background staining
- Available in a variety of colors
Specifications
Specifications
| Frosted | Yes |
| Type | Microscope Slide |
| Material | Glass |
| Color | Blue |
| Quantity | 100/Cs. |