Learn More
Epredia™ Shandon™ ColorFrost™ Plus Slides
Catalog No. 9991004 Shop All Epredia Products
Change view
Click to view available options
Color:
Aqua
Blue
Burnt Orange
Gold
Green
Lavender
Pink
Tan
Yellow
Quantity:
100/Cs.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
99-910-04 Pink 100/Cs.
99-910-01 Blue 100/Cs.
99-910-02 Yellow 100/Cs.
99-910-03 Green 100/Cs.
99-910-09 Lavender 100/Cs.
99-910-11 Burnt Orange 100/Cs.
99-910-12 Tan 100/Cs.
99-910-13 Aqua 100/Cs.
99-910-14 Gold 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 99-910-04 Supplier Epredia™ Supplier No. 9991004
Add to Cart
Add to Cart
This item is not returnable. View return policy
Epredia™ Shandon ColorFrost Plus Slides are positively charged for improved cellular adhesion.
  • Electrostatically attract tissue sections and cytology preparations, binding them to the slide
  • Eliminate the need for special adhesives or protein coatings
  • No background staining
  • Available in a variety of colors
Specifications
Frosted Yes
Type Microscope Slide
Material Glass
Color Pink
Quantity 100/Cs.