Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Shandon™ Dissecting Tray for Raised Dissecting Boards, 12x 11 in.

Catalog No. 3611403 Shop All Epredia Products
Change view
Click to view available options
Length (English):
12 in.
23 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
3611403 23 in.
35-614 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3611403 Supplier Epredia™ Supplier No. 3611403
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Epredia™ Shandon Dissecting Tray for Raised Dissecting Boards feature ample space for drainage and instruments.

  • Four embossments on tray bottom hold Raised Dissecting Boards 356 and 35614

Specifications

Product Type Dissecting Tray
Material Polyethylene
Size Large
Length (English) 23 in.
Width (English) 16 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.