Description
- Designed to hold tissue specimens during processing and embedding
- Acetal polymer cassettes with writing face at 45° angle
- Pore style: Slotted
- Locking lid attached by two hinges
Specifications
Specifications
| Quantity | 25 Cassettes x 30 Tubes |
| Color | Green |