Description
- Acetal polymer cassettes with writing face at 45° angle
- Slotted pore style
- Locking lid attached by two hinges
Ordering Information:
- Tube packs are supplied in 10 tubes of 75 cassettes each
Specifications
Specifications
| Pore Style | Slotted Pore |
| Writing Face Angle | 45° |
| Color | Blue |
| Material | Acetal Polymer |
| Product Type | Biopsy Cassette |