Learn More
Epredia™ Shandon™ Embedding Cassettes II in Tube Packs, aqua
Catalog No. B1000729LI Shop All Epredia Products
Change view
Click to view available options
Color:
Blue
Lilac
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
B1000729LI Lilac
B1000729BL Blue
B1000729WH White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. B1000729LI Supplier Epredia™ Supplier No. B1000729LI
This item is not returnable. View return policy
Hold tissue specimens during processing and embedding with Epredia™ Shandon™ Embedding Cassettes II in Tube Packs.
  • Acetal polymer cassettes with writing face at 45° angle
  • Slotted pore style
  • Locking lid attached by two hinges
Ordering Information:
  • Tube packs are supplied in 10 tubes of 75 cassettes each
Specifications
Pore Style Slotted Pore
Writing Face Angle 45°
Color Lilac
Material Acetal Polymer
Product Type Biopsy Cassette