Description
- Can be used on all Epredia PrintMate automatic cassette labelers
- Fit securely in microtome chuck adapters
- Designed to accept metal lids (Order number 1001003, 25/pk., sold separately)
- Made of acetal polymer with a pore style of round holes
- Supplied in 10 tubes of 75 cassettes each
Specifications
Specifications
| Pore Style | Round Holes |
| Quantity | 750 Cassettes/Cs. |
| Color | Gray |
| Material | Acetal Polymer |