Learn More
Epredia™ Shandon™ Eosin-Y Stain
Catalog No. 6766010 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1L
4L
Each
Product Type:
Alcoholic Eosin Y Stain
Aqueous Eosin Y Stain
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
6766010 Each Aqueous Eosin Y Stain
6766007 1L Alcoholic Eosin Y Stain
6766008 4L Alcoholic Eosin Y Stain
6766009 1L Aqueous Eosin Y Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6766010 Supplier Epredia™ Supplier No. 6766010
Add to Cart
Add to Cart
Produce brilliant staining with Epredia™ Shandon™ Eosin-Y, a fast-acting cytoplasmic counterstain.
  • Ready-to-use
  • Produces consistent results
  • Stable and precipitate-free; no filtering
  • Compatible with all alum hematoxylin stains
  • Available in alcoholic or aqueous formulations
Specifications
Quantity Each
Product Type Aqueous Eosin Y Stain
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov