Learn More
Epredia™ EZ Double Cytofunnel™
Catalog No. A78710021 Shop All Epredia Products
Change view
Click to view available options
Quantity:
200/Cs.
40 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A78710021 200/Cs.
A78710005 40 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. A78710021 Supplier Epredia™ Supplier No. A78710021
Add to Cart
Add to Cart
This item is not returnable. View return policy
Double the number of sample presentations per run with the Epredia™ EZ Double Cytofunnel™.
  • Disposable sample chambers with white filter cards and caps
  • Funnel markings corresponding to slide markings for simple identification
  • Pre-attached filter card
Specifications
Color White
Disposable Single-use
Quantity 200/Cs.
Type EZ Double Cytofunnel™, With Filter Cards and Caps
Volume (Metric) 0.05 to 0.25 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov