Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Glyo-Fixx™

Catalog No. 9990924 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
125 x 60mL
180 x 20mL
5 gal.
125 x 120mL
Product Type:
Fixative
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
9990924 125 x 60mL Fixative
6764260 5 gal. Fixative
67-642-62 1 gal. Fixative
9990928 125 x 120mL Fixative
9990920 180 x 20mL Fixative
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 9990924 Supplier Epredia™ Supplier No. 9990924
This item is not returnable. View return policy

Obtain superior results to neutral buffered formalin with Glyo-Fixx™, a noncarcinogenic, nonsensitizing, and environmentally friendly substitute.

  • Produces distinct and crisp cellular morphology
  • Fixes most antigens without impairing immunoreactivity
  • Allows special stain dyes, including silver procedures, to be sharp without background staining
  • Presents fine detail in specimen structure

Specifications

Product Type Fixative
Quantity 125 x 60mL
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.