Learn More
Epredia™ Shandon™ Grosslab™ Filter Sets
Catalog No. 1001618 Shop All Epredia Products
Change view
Click to view available options
For Use With (Equipment):
Grosslab Junior Series 9600
Grosslab Senior Series 8700 and 9700
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
10-016-18 Grosslab Junior Series 9600
23-245-560 Grosslab Senior Series 8700 and 9700
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-016-18 Supplier Epredia™ Supplier No. 1001618
Add to Cart
Edge
Add to Cart
Each filter set contains a formaldehyde neutralizing (Potassium Permanganate) filter along with a fine particle (Charcoal) filter suitable for Grosslab Jr. and Grosslab Sr. workstations.
  • Stand with extruded aluminum frame and stainless-steel shelf has an adjustable back panel (stand is height adjustable from 28 to 42 in. [71.1 to 106.7cm ])
  • Stand-alone dictation systems include a handheld microphone and power supply
  • Towel bar for rolled towels requires Towel Dispenser (10-702-14)
Specifications
Product Type Filter Set
Description Filter Size: 48.3 cm x 19.7 cm x 5.7 cm
For Use With (Equipment) Grosslab Junior Series 9600