Learn More
Epredia™ Shandon™ Surgical Lighting
Epredia™ Shandon™ Surgical Lighting is available in a variety of configurations.
Supplier:  Epredia™ 20580
 View more versions of this product
Catalog No. 20580
Description
Description
  • Complete line of lights provides surgical-quality lighting for use in applications where high intensities and color correction are needed, but field size requirements are less critical, such as in pathology and animal surgery applications
Specifications
Specifications
Track-mount, dual head
Surgical Lighting
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.