Learn More
Epredia™ Xylene Substitute
Catalog No. 9990507 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
1.32 gal.
5 gal.
4 x 1 gal.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9990507 5 gal.
6764506 4 x 1 gal.
9990505 1 gal.
67-645-15 1.32 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 9990507 Supplier Epredia™ Supplier No. 9990507
Add to Cart
Add to Cart
This item is not returnable. View return policy
Achieve safer, more convenient processing, staining and coverslipping with Epredia™ Xylene Substitute.
  • Not sensitizing to normal skin
  • Evaporation rate similar to xylene
  • Virtually odorless aliphatic hydrocarbon—no citrus aroma
  • Airborne exposure limit three times safer than xylene (TLV 300ppm)
Specifications
Quantity 5 gal.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov