Learn More
Thomas Scientific TIPS, 10UL LOW RETENTION, SHARP FILTER/BARRIER
SDP
Supplier:  Thomas Scientific 1158U34
Encompass_Preferred
SHARP precision barrier tips eliminate false signals and cross contamination caused by aerosols. SHARP tips feature Denville's new low retention polymer technology that virtually eliminates sample retention. Our inert hydrophobic barrier prevents pipettor contamination without trapping liquid or contaminating samples. These tips provide the most accurate sample delivery in the industry. They contain no nucleic acids, PCR inhibitors, detectable nucleases or endotoxins. All sizes are natural color, presterilized and free of metallic content.
Catalog No. NC1609458