Learn More
Sterilis Solutions Sharps Container, 4 Gallon Cylinder

Description
- General-purpose sharps container with clear locking lid and marked with biohazard label
- Compliant with FDA and OSHA standards for sharps and biohazard waste collection
- Puncture-resistant with wide circular opening for vertical or horizontal loading of needles
- Made from transparent red puncture-resistant polypropylene that can be autoclaved or incinerated
- Rotating handle for easy transport and handling
- Nests to conserve storage space
Specifications
Specifications
| Capacity (Metric) | 15 L |
| Capacity (English) | 4 gal. |
| Color | Translucent Red |
| Dimensions (L x W) | 11.37 x 13 in. |
| Disposable | Single-use |
| Height (Metric) | 330.1 mm |
| Latex Free | Yes |
| Quantity | 40 Pk. |
| Shape | Cylindrical |
| Stackable | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.