Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Sharps Container

Catalog No. 22730456 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English):
1 qt.
1.5 qt.
2.2 qt.
1 gal.
5 qt.
5.4 qt.
2 gal.
3 gal.
3.5 gal.
7 gal.
8 gal.
Closure Type:
Chimney Top Lid
Flip Top Lid
Horizontal Drop Lid
Mailbox Lid
Rotating Lid
Rotating and Split Lid
Sliding Lid
Split Lid
Dimensions (L x W x H):
10 x 4.4 x 9.2 in.
10 x 4.4 x 9.7 in.
10 x 7 x 14 in.
10.2 x 6.9 x 5.6 in.
10.25 x 7 x 9.75 in.
10.3 x 6.8 x 10.1 in.
10.3 x 7 x 5.8 in.
10.3 x 7 x 9.7 in.
11.3 x 7 x 13.3 in.
11.3 x 7 x 7.3 in.
12.1 x 4.7 x 10 in.
13.4 x 6 x 13.5 in.
13.4 x 6 x 13.6 in.
15.3 x 10.9 x 17.3 in.
15.40 x 11.89 x 13.79 in.
3.9 x 3.8 x 6.1 in.
4.5 x 4.5 x 6.6 in.
4.7 x 4.0 x 9.1 in.
6.3 x 4.6 x 6.7 in.
Opening Size (English):
1.6 x 1.25 in.
1.9 x 1.4 in.
1.9 x 1.7 in.
2.5 x 2.5 in.
3.4 x 1.5 in.
4 x 1.8 in.
5.5 x 2 in.
8 x 2.8 in.
8.2 x 2
8.5 x 1.5 in.
8.5 x 1.8 in.
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Closure Type Dimensions (L x W x H) Opening Size (English)
22-730-456 1.5 qt. Flip Top Lid 4.7 x 4.0 x 9.1 in. 1.9 x 1.7 in.
22-730-455 1 qt. Sliding Lid 4.5 x 4.5 x 6.6 in. 1.9 x 1.4 in.
22-730-457 1 qt. Flip Top Lid 3.9 x 3.8 x 6.1 in. 1.6 x 1.25 in.
22-730-458 2.2 qt. Rotating Lid 6.3 x 4.6 x 6.7 in. 3.4 x 1.5 in.
22-730-459 5.4 qt. Flip Top Lid 12.1 x 4.7 x 10 in. 8 x 2.8 in.
22-730-460 5 qt. Horizontal Drop Lid 10 x 4.4 x 9.7 in. 8.5 x 1.5 in.
22-730-461 5 qt. Mailbox Lid 10 x 4.4 x 9.2 in. 8.5 x 1.5 in.
22-730-462 1 gal. Sliding Lid 10.3 x 7 x 5.8 in. 5.5 x 2 in.
22-730-463 1 gal. Chimney Top Lid 11.3 x 7 x 7.3 in. 4 x 1.8 in.
22-730-464 1 gal. Rotating Lid 10.2 x 6.9 x 5.6 in. 4 x 1.8 in.
22-730-465 2 gal. Sliding Lid 10.3 x 6.8 x 10.1 in. 5.5 x 2 in.
22-730-466 2 gal. Chimney Top Lid 11.3 x 7 x 13.3 in. 4 x 1.8 in.
22-730-467 2 gal. Flip Top Lid 10.3 x 7 x 9.7 in. 8.2 x 2
22-730-468 2 gal. Rotating Lid 10.25 x 7 x 9.75 in. 4 x 1.8 in.
22-730-469 3 gal. Sliding Lid 13.4 x 6 x 13.6 in. 2.5 x 2.5 in.
22-730-470 3.5 gal. Chimney Top Lid 10 x 7 x 14 in. 4 x 1.8 in.
22-730-471 3 gal. Rotating and Split Lid 13.4 x 6 x 13.5 in. 4 x 1.8 in.
22-730-472 7 gal. Split Lid 15.40 x 11.89 x 13.79 in. 8.5 x 1.8 in.
22-730-473 8 gal. Rotating and Split Lid 15.3 x 10.9 x 17.3 in. 4 x 1.8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
This item is not returnable. View return policy
Catalog No. 22-730-456
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Fisherbrand Sharps Container, designed for higher volume usage in laboratories, doctors' offices, and clinics for larger biohazardous waste items, enables laboratory or medical professionals to temporarily seal the container for secure internal transportation.

  • Provides safe disposal of needles and syringes.
  • The container is available in various sizes to accommodate different requirements, and it features different lid designs to suit various preferences.

Specifications

Certifications/Compliance ISO 23907, OSHA 29 CFR 1910.1030, FDA 510k
Depth (English) 9.1 in.
Closure Type Flip Top Lid
Closure Color Clear, White
Capacity (English) 1.5 qt.
Color Red
Dimensions (L x W x H) 4.7 x 4.0 x 9.1 in.
Disposable Single-use
Latex Free Yes
Opening Size (English) 1.9 x 1.7 in.
Quantity 36/Cs
Shape Rectangle
Stackable Yes
Sterility Non-sterile
Autoclavable Autoclavable
Width (English) 4.0 in.
Length (English) 4.7 in.
Height (English) 9.1 in.
Packaging Case
Material Polypropylene
Product Type Sharps Disposal Container
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.