Learn More
Oak Ridge Sharps Container with Rotary Lid
Catalog No. 22730448
Change view
Click to view available options
Capacity (Metric):
3.7 L
7.5 L
11.3 L
30.2 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22-730-448 7.5 L
22-730-445 3.7 L
22-730-450 11.3 L
22-730-451 30.2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-730-448 Supplier Oak Ridge Supplier No. 0320150R
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed for multi purpose use
  • Designed for Multi purpose use
  • Rotary Lid maximizes usable container volume for larger sharps
  • Shatter and puncture-resist
  • Locks for final disposal
Specifications
Capacity (Metric) 7.5 L
Closure Type Rotary lid
Capacity (English) 2 gal.
Material Polypropylene
Product Type Sharps Container