Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Shatterproof Plastic Ruler
SCI_ED

Catalog No. S43612 Shop All Eisco Products
Change view
Click to view available options
Length (English):
6 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
S43612 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43612 Supplier Eisco™ Supplier No. PH201703
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for student use.

  • 6 in. (15cm) ruler
  • Clear with black markings
  • With inch and cm markings

Specifications

Product Type Ruler
Length (English) 6 in.
Length (Metric) 15 cm
Material Shatterproof Plastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.