Learn More
Ergodyne™ SHAX™ 6098 Pop-Up Tent Sidewalls
Catalog No. 19819716 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Blue
Lime
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-716 Lime
19-819-715 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-819-716 Supplier Ergodyne™ Supplier No. 12998
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Offers privacy and protection from the sun, wind and rain
  • Compliments all sides of the SHAX™ 6000 and 6010 Pop-Up Tents, or the 10 ft side of the SHAX™ 6015 Pop-Up Tent
  • Fire-retardant panel is made with heavy-duty 300-denier polyester fabric with waterproofing polyurethane coating (PU) and UV treatments for added protection.
  • Easy-to-use hook and loops straps securely attach the tent wall to the frame
  • Wind-resistant fabric
  • Packs up easily
  • Reinforced on the corners for added durability
  • hH-vis lime option and silver coating provide increased visibility on busy job sites or at events
Specifications
Color Lime
Dimensions (L x W) 10 x 10 ft. (3 x 3 m)
Quantity 1/Cs.