Learn More
Enzo Life Sciences Shc monoclonal antibody (pSer36) (6E10) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804358C100
Encompass_Preferred
Clone: 6E10. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 33-39 (E33LPpSPSA39) of shc phosphorylated at Ser36 conjugated to KLH. Species reactivity: Human, Mouse, Dog. Applications: WB. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-200-9883
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.