Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101882
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101882 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101882 Supplier Invitrogen™ Supplier No. PA5101882
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SHD.

May function as an adapter protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHD
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SHD
Gene Accession No. Q96IW2
Gene Alias AI413439; SH2 domain-containing adapter protein D; Shd; Src homology 2 domain containing transforming protein D; src homology 2 domain-containing transforming protein D
Gene Symbols SHD
Host Species Rabbit
Immunogen A synthesized peptide derived from human SHD(Accession Q96IW2), corresponding to amino acid residues G140-D190.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56961
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.