Learn More
Grainger Shear 6.5 A Double Cut 2 500 spm
Supplier:  Grainger 14F138
Encompass_Preferred
The Grainger 6.5 Amp Double Cut Shear has a 14 gauge capacity for mild steel. It can make straight cuts up to 16" and has a cutting speed of 500 strokes per minute. The overall dimensions are 20" x 7" x 7.5" and it weighs 15 lbs. This electric metal shear features a foot pedal operation and has a 6.5 amp motor. It's designed for straight cuts in mild steel plate, sheet metal and small solids.
Catalog No. 50-275-3501