Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sheep anti-Bovine IgA Heavy Chain Secondary Antibody [Janelia Fluor 525], R&D Systems™
SDP

Catalog No. NB782273 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB782273 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB782273 Supplier R&D Systems Supplier No. NBP246599JF525
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Sheep Polyclonal Antibody

IgA Heavy Chain Polyclonal antibody specifically detects IgA Heavy Chain in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen IgA Heavy Chain
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Host Species Sheep
Immunogen Bovine IgA
Purification Method Immunogen affinity purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.