Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sheep anti-Bovine IgA Secondary Antibody, Janelia Fluor 549, Novus Biologicals™
SDP

Catalog No. NB555744 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB555744 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB555744 Supplier Novus Biologicals Supplier No. NBP268347JF549
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

IgA Polyclonal specifically detects IgA in Bovine samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.

Specifications

Antigen IgA
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias Immunoglobulin A
Host Species Sheep
Immunogen Purified bovine IgA
Purification Method Affinity purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3493
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.