Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sheep anti-Bovine IgG (H+L) Secondary Antibody, R&D Systems™
SDP

Catalog No. NB775 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB775 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB775 Supplier R&D Systems Supplier No. NB775
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Sheep Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Bovine samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, ELISA, Immunohistochemistry 1:200 - 1:2000, Immunocytochemistry/Immunofluorescence 1:200 - 1:2000, Immunohistochemistry-Paraffin
Host Species Sheep
Immunogen Bovine IgG-heavy and light chain
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity By immunoelectrophoresis and ELISA this reacts specifically with bovine IgG and with light chains common to other bovine immunoglobulins. No was detected against non-immunoglobulin serum proteins. This may cross react with IgG from other species.
Target Species Bovine
Content And Storage Store at 4C. Do not freeze.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.