Learn More
Sheep anti-Bovine IgG Heavy Chain Secondary Antibody, DyLight 405, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgG Heavy Chain |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 405 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Immunoglobulin G, ImmunoglobulinG |
| Host Species | Sheep |
| Immunogen | Bovine IgG-heavy chain |
| Purification Method | Immunogen affinity purified |
| Quantity | 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.