Learn More
Description
Specifications
Specifications
| Antigen | IgG Heavy Chain |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | HRP |
| Dilution | Western Blot 1:5000 - 1:50000, ELISA 1:10000-1:100000, Immunohistochemistry 1:200 - 1:5000, Immunocytochemistry/Immunofluorescence 1:200 - 1:5000, Immunohistochemistry-Paraffin 1:200 - 1:5000 |
| Gene Alias | Immunoglobulin G, ImmunoglobulinG |
| Host Species | Sheep |
| Immunogen | Bovine IgG-heavy chain |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
