Learn More
Sheep anti-Bovine IgG1 Heavy Chain Secondary Antibody, DyLight 594, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgG1 Heavy Chain |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Host Species | Sheep |
| Immunogen | Bovine IgG1 |
| Purification Method | Immunogen affinity purified |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.