Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sheep anti-Bovine IgG1 Heavy Chain Secondary Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB783 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB783 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB783 Supplier R&D Systems Supplier No. NB783
Add to Cart
Add to Cart

Sheep Polyclonal Antibody has been used in 13 publications

IgG1 Heavy Chain Polyclonal specifically detects IgG1 Heavy Chain in Bovine samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen IgG1 Heavy Chain
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate HRP
Dilution Western Blot 1:2000-1:20000, ELISA 1:10000-1:100000, Immunohistochemistry 1:200 - 1:2000, Immunocytochemistry/Immunofluorescence 1:200 - 1:2000, Immunohistochemistry-Paraffin
Host Species Sheep
Immunogen Bovine IgG1
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity By immunoelectrophoresis and ELISA this reacts specifically with bovine IgG1. This may cross react with IgG1 from other species.
Target Species Bovine
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.