Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sheep anti-Bovine IgG1 Secondary Antibody [Janelia Fluor 669], Novus Biologicals™
SDP

Catalog No. NB783450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB783450 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB783450 Supplier R&D Systems Supplier No. NBP268328JF669

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Sheep Polyclonal Antibody

IgG1 Polyclonal antibody specifically detects IgG1 in Bovine samples. It is validated for ELISA, Immunodiffusion

Specifications

Antigen IgG1
Applications ELISA, Immunodiffusion
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias IgG1, Igh-4, RGD1359539, VH7183
Host Species Sheep
Immunogen Purified bovine IgG1
Purification Method Affinity purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.