Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sheep anti-Bovine IgG2 Secondary Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB554618 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB554618 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB554618 Supplier Novus Biologicals Supplier No. NBP268333B
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

IgG2 Polyclonal specifically detects IgG2 in Bovine samples. It is validated for ELISA, Immunodiffusion.

Specifications

Antigen IgG2
Applications ELISA, Immunodiffusion
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias Constant region of heavy chain of IgG2, immunoglobulin gamma 2 (Gm marker), immunoglobulin heavy constant gamma 2 (Gm marker)
Host Species Sheep
Immunogen Purified bovine IgG2
Purification Method Affinity purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3501
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.