Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sheep anti-Bovine IgM (H+L) Secondary Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB552208 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB552208 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB552208 Supplier R&D Systems Supplier No. NBP268322R

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Sheep Polyclonal Antibody

IgM (H+L) Polyclonal specifically detects IgM (H+L) in Bovine samples. It is validated for ELISA, Immunodiffusion.

Specifications

Antigen IgM (H+L)
Applications ELISA, Immunodiffusion
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Host Species Sheep
Immunogen Purified bovine IgM
Purification Method Affinity purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.