Learn More
Sheep anti-Bovine IgM (H+L) Secondary Antibody, Janelia Fluor 549, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgM (H+L) |
| Applications | ELISA, Immunodiffusion |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 549 |
| Formulation | 50mM Sodium Borate |
| Host Species | Sheep |
| Immunogen | Purified bovine IgM |
| Purification Method | Affinity purified |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.