Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sheep anti-Canine IgG2 Secondary Antibody, Janelia Fluor 646, R&D Systems™
SDP

Catalog No. NB554051 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB554051 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB554051 Supplier R&D Systems Supplier No. NBP268359JF646
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Sheep Polyclonal Antibody

IgG2 Polyclonal specifically detects IgG2 in Canine samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.

Specifications

Antigen IgG2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Constant region of heavy chain of IgG2, immunoglobulin gamma 2 (Gm marker), immunoglobulin heavy constant gamma 2 (Gm marker)
Host Species Sheep
Immunogen Canine IgG2
Purification Method Affinity purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3501
Target Species Canine
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.