Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sheep anti-Guinea Pig IgM Fc Secondary Antibody, R&D Systems™
SDP

Catalog No. NBP269694 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-69694 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-69694 Supplier R&D Systems Supplier No. NBP269694
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Sheep Polyclonal Antibody

IgM Fc Polyclonal specifically detects IgM Fc in Guinea Pig samples. It is validated for ELISA, Immunoelectrophoresis, Immunoprecipitation.

Specifications

Antigen IgM Fc
Applications ELISA, Immunoelectrophoresis, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunoprecipitation 1:10 - 1:500
Formulation Whole antiserum with No Preservative
Host Species Sheep
Immunogen Highly purified normal IgM isolated from pooled guinea pig serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Whole Antiserum
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity The reactivity of the antiserum is restricted to the Fc part of the IgM molecule. In immunoelectrophoresis and radial immunodiffusion, using various antiserum concentrations against normal guinea pig serum a single precipitin line is obtained which shows a reaction of identity with the precipitin line obtained with purified IgM. No precipitation reaction is obtained with purified IgG, IgA, and IgG/Fab fragments.
Reconstitution Reconstitute the lyophilized antiserum by adding 1 ml sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2).
Target Species Guinea Pig
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Lyophilized
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.