Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sheep anti-Human Lambda Light Chain Secondary Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB538977 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB538977 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB538977 Supplier Novus Biologicals Supplier No. NB7482G
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Lambda Light Chain Polyclonal specifically detects Lambda Light Chain in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Lambda Light Chain
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias IGLC, immunoglobulin lambda constant group
Host Species Sheep
Immunogen Human Lambda Free Light Chain
Purification Method Immunogen affinity purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3537
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.