Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sheep anti-Rabbit IgG (H+L) Secondary Antibody, Texas Red (Pre-adsorbed), Novus Biologicals™
SDP

Catalog No. NBP173667 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP173667 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP173667 Supplier R&D Systems Supplier No. NBP173667

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Sheep Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Flow Cytometry, Fluorophore-linked immunosorbent assay, Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen IgG (H+L)
Applications Flow Cytometry, Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Texas Red
Dilution Western Blot 1:1000 - 1:10000, Flow Cytometry 1:500 - 1:2500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000
Host Species Sheep
Immunogen Rabbit IgG whole molecule
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity This antibody was pre-adsorbed against Bovine, Chicken, Goat, Guinea Pig, Horse, Human, Mouse, Rat, and Sheep Serum Proteins. No reaction was observed against Bovine, Chicken, Goat, Guinea Pig, Horse, Human, Mouse, Rat and Sheep Serum Proteins.
Reconstitution Reconstitute with 1.0 ml deionized water (or equivalent).
Target Species Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.