Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sheep anti-Rat IgG (H+L) Secondary Antibody, HRP, R&D Systems™
SDP

Catalog No. NBP222109 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222109 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222109 Supplier R&D Systems Supplier No. NBP222109
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Sheep Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rat samples. It is validated for ELISA, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 2.0 mg/mL
Conjugate HRP
Dilution Western Blot 1:1000-1:10000, ELISA 1:5000-1:100000, Immunohistochemistry 1:500-1:2500
Formulation PBS, pH 7.2 with 10mg/mL BSA (IgG and protease free) with 0.01% Gentamicin Sulfate
Gene Gene ID 3500
Gene Alias IgG, immunoglobulin heavy constant epsilon
Gene Symbols IGHG1
Host Species Sheep
Immunogen Purified rat IgG.
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity Recognizes rat IgG. Assay by Immunoelectrophoresis resulted in a single precipitin arc against anti-peroxidase, anti-sheep serum, rat IgG and rat serum.
Target Species Rat
Content And Storage Store at 4C in the dark.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.