Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Sheep anti-Rat IgG (H+L) Secondary Antibody, HRP
Sheep Polyclonal Secondary Antibody
Supplier: Invitrogen™ PA128642
Description
PA1-28642 detects Rat IgG from rat samples.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Rat IgG (H+L) | |
Polyclonal | |
HRP | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
Rat | |
Secondary Antibody | |
IgG |
ELISA, Immunohistochemistry, Western Blot | |
2 mg/mL | |
PBS with 1% BSA and 0.01% gentamicin sulfate; pH 7.2 | |
Rat IgG whole molecule. | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction