Learn More
Shelby™ Specialty 5280 Gloves
Catalog No. 19167782 Shop All Shelby Specialty Gloves Products
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-167-782 Small
19-167-783 Medium
19-167-780 2X-Small
19-167-781 X-Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-167-782 Supplier Shelby™ Supplier No. 5280GS
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Specialty thermal gloves are resistant to flames, heat, and punctures for fire management or other industrial applications.
  • Heavyweight, gauntlet-style gloves
  • Grain elkhide leather along with Shelby abrasion-resistant brushed pigskin leather
  • Shelby Black Line Nomex™ 101.6 mm (4 in.) wristlet
  • GORE RT7100 glove barrier fabric
  • Gunn-cut pattern with wing thumb and wrap-around index finger
Certifications
SEI Certified
Specifications
Certifications/Compliance NFPA 1971, CE PPE, EN 659:1996, EN 388:1994
Color Tan
Material Pigskin Leather, Elkskin Leather
Product Type Structural Firefighting Gloves
Size Small
Glove Cuff Style Gauntlet
Liner Material Thermal