Learn More
Thermo Scientific™ Shelf Kits for Thermo Scientific™ Ultra-Low Temperature Freezers
Catalog No. SK600
Change view
Click to view available options
For Use With (Equipment):
Forma 88300; TSU300; HFU300T; Revco UxF300
Forma 88400; TSU400; HFU400T; Revco UxF400
Forma 88500; TSU500; HFU500T; Revco UxF500
Forma 88600; TSU600; HFU600T; Revco UxF600
Forma 88700; TSU700; HFU700T; Revco UxF700
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
SK600 Forma 88600; TSU600; HFU600T; Revco UxF600
SK300 Forma 88300; TSU300; HFU300T; Revco UxF300
SK400 Forma 88400; TSU400; HFU400T; Revco UxF400
SK500 Forma 88500; TSU500; HFU500T; Revco UxF500
SK700 Forma 88700; TSU700; HFU700T; Revco UxF700
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. SK600 Supplier Thermo Scientific™ Supplier No. SK600
Add to Cart
Add to Cart
Maximize storage space with shelf kits designed for Thermo Scientific™ Ultra-Low Temperature Freezers.
Maximize storage space with Thermo Scientific™ Shelf Kits for Thermo Scientific™ Ultra-Low Temperature Freezers; designed for use with with Thermo Scientific™ Forma™ 8800, HERAfreeze™ HFU-T, Revco™ UxF, and TSU Series freezers.
TRUSTED_SUSTAINABILITY
Specifications
Description Includes one shelf and clips
Type Shelf Kit
Shelves 1
For Use With (Equipment) Forma 88600; TSU600; HFU600T; Revco UxF600