Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Shelves for Fisherbrand™ Isotemp™ Freezers, Refrigerators, and Refrigerator/Freezers

Catalog No. 13986210B Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
127G, 227R, 227G Isotemp
133G, 233G Isotemp
145G, 245G Isotemp
149G, 172G, 249R, 249G, 272G Isotemp
Isotemp 23, 47, 74 cu. ft.
Isotemp 45 cu. ft.
Models 525D, Model 525F
Type:
Extra Support Clips
Full Shelf
Half Shelf
Shelf
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Type
13-986-210B 133G, 233G Isotemp Full Shelf
13-987-70 Models 525D, Model 525F Extra Support Clips
13-986-210H 145G, 245G Isotemp Half Shelf
13-986-210A 127G, 227R, 227G Isotemp Full Shelf
13-986-210D 149G, 172G, 249R, 249G, 272G Isotemp Full Shelf
13-986-210E Isotemp 23, 47, 74 cu. ft. Full Shelf
13-986-210J 149G, 172G, 249R, 249G, 272G Isotemp Half Shelf
13-986-210L Isotemp 45 cu. ft. Shelf
13-986-211E Isotemp 23, 47, 74 cu. ft. Full Shelf
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 13-986-210B

Epoxy coated steel for easy cleaning.

TRUSTED_SUSTAINABILITY

Specifications

Type Full Shelf
Cabinet Material Steel
Exterior Finish Epoxy
Color White
For Use With (Equipment) 133G, 233G Isotemp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.