Learn More
Fisherbrand™ Shelves for Fisherbrand™ Isotemp™ Freezers, Refrigerators, and Refrigerator/Freezers
Shop All Fisherbrand ProductsSpecifications
Specifications
| Type | Half Shelf |
| Cabinet Material | Epoxy-coated |
| Exterior Finish | Epoxy |
| Color | White |
| For Use With (Equipment) | 145G, 245G Isotemp |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.