Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Shiga like-toxin 1 Monoclonal Antibody (13C4)
GREENER_CHOICE

Catalog No. 01676396 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-396 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-396 Supplier Invitrogen™ Supplier No. MA140277
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-40277 detects Shigatoxin 1 from E. coli samples. MA1-40277 has been successfully used in ELISA, functional assays and Western blot applications. The MA1-40277 immunogen is 1B subunit of Shigatoxin 1.

Shiga-like toxins (SLTs), are also called Verotoxins. Enterohemorrhagic Escherichia coli (EHEC) strains which are primarily of serotypes 0157:H7, 026:H11 and O111:H8 have been incriminated as etiologic agents of hemorrhagic colitis and Hemolytic-uremic syndrome, a generalized disease characterized by acute renal failure, thrombocytopenia, and microangiopathic hemolytic anemia. There are several distinct E.coli SLTs. SLT-I and SLT-II are produced by EHEC. SLT-I and Shiga toxin share 99% deduced amino acid sequence homology, whereas SLT-I and SLT-II share about 60% deduced amino acid sequence homology. SLT-I and SLT-II are antigenically distinct. The protein structure of the toxin consists of two domains: the A polypeptide that inhibits protein synthesis by targeting ribosomes, and the B polypeptide pentamer that binds to the eukaryotic cell receptor globotriaosylceramide (Gb3) leading to receptor-mediated endocytosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Shiga like-toxin 1
Applications ELISA, Functional Assay, Western Blot
Classification Monoclonal
Clone 13C4
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and no preservative
Gene Alias SLT1; SLT-1; SLTI; SLT-I; Verocytotoxin; Verotoxin
Host Species Mouse
Immunogen 1B subunit of Shigatoxin 1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.