Learn More
Invitrogen™ Shiga toxin 2A Monoclonal Antibody (11E10)

Description
Product is supplied at 1 mL (100 μL/mL). Application Notes: Functional assay - For inhibition of biological activity in vitro dilutions have to be made according to the amounts of toxin to be inactivated. ELISA - antibody can be used as detector.
Specifications
Specifications
| Antigen | Shiga toxin 2A |
| Applications | ELISA, Functional Assay, Western Blot |
| Classification | Monoclonal |
| Clone | 11E10 |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and no preservative |
| Gene Alias | Shiga toxin; Shigatoxin Type 2; Shigatoxin Type II; STX2; STX-2a; STX-IIa |
| Host Species | Mouse |
| Purification Method | Protein G |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.