Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Shigella boydii Monoclonal Antibody (32/32)
GREENER_CHOICE

Catalog No. PIMA546610
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546610 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA546610 Supplier Invitrogen™ Supplier No. MA546610
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Shigella boydii Monoclonal Antibody for ICC/IF, ELISA

Shigellae are Gram-negative, nonmotile, facultatively anaerobic, non-spore-forming rods. Shigella are differentiated from the closely related Escherichia coli on the basis of pathogenicity, physiology (failure to ferment lactose or decarboxylate lysine) and serology. The genus is divided into four serogroups with multiple serotypes: A (S dysenteriae, 12 serotypes); B (S flexneri, 6 serotypes); C (S boydii, 18 serotypes); and D (S sonnei, 1 serotype). [PMID: 21413292].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Shigella boydii
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 32/32
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Alias S. boydii; Shigella
Host Species Mouse
Immunogen NCTC 9731.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.