Learn More
Bioss Treponema pallidum
SDP
Supplier:  Bioss BSM72445M
Encompass
Treponema pallidum Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications WB ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-65536
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.