Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Shisa-4 Recombinant Protein Antigen
SDP

Catalog No. NB171251 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171251 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171251 Supplier Novus Biologicals™ Supplier No. NBP317436PEP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 149345
Molecular Weight (g/mol) M.W. Theoretical: 21 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias C1orf40, chromosome 1 open reading frame 40, FLJ18233, hShisa4, MGC129812, MGC129813, protein shisa-4, shisa homolog 4 (Xenopus laevis), TMEM58, Transmembrane protein 58MGC102893
Gene Symbol SHISA4
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein Shisa-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.