Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Shoe Cover Holder
SDP

Catalog No. 01259210
Change view
Click to view available options
Mounting:
Magnetic Mount
VHB Tape
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
01-259-210 Magnetic Mount
01-259-162 VHB Tape
01-259-144 Wall Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
N/A
Catalog No. 01-259-210 Supplier Poltex Supplier No. BOOTY1M
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N/A

Holds and dispenses shoe covers or equivalent. Choose from Wall Mount, HB Tape Mount, or Magnet Mount with durable and cleanable PETG construction.

  • Holds and dispenses X-Tra Traction Shoe Covers or equivalent
  • Available in Wall Mount (screws and anchors included)
  • Available in HB Tape Mount (high-bond tape included)
  • Available in Magnet Mount (powerful magnets included)
  • Durable and cleanable PETG construction
  • Consumables not included

Specifications

Product Type Shoe Cover Dispenser
Color Clear
Dimensions (L x W x H) 5.8 x 10.4 x 12.3 in.
Holds Shoe Covers, PPE, Bouffant Caps
Material PETG
Mounting Magnetic Mount
No. of Compartments 1
Product Style Open Top and Front
Edges Rounded
Formulation PETG
Includes Magnets for Mounting
Quantity 1 Ea.
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.